Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191745.1 | NP_001178674.1 | 9 - 10 | 1362 | 64 | |
XM_008762509.1 | XP_008760731.1 | 9 - 10 | 1616 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |