Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcea2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057098.1 | NP_476439.1 | 5 - 6 | 539 | 89 | |
XM_006235740.1 | XP_006235802.1 | 5 - 6 | 631 | 89 | ||
XM_006235741.1 | XP_006235803.1 | 5 - 6 | 616 | 89 | ||
XM_006235742.2 | XP_006235804.1 | 5 - 6 | 605 | 89 | ||
GenBank mRNA | D12927.1 | - | 5 - 6 | 539 | 89 | |
FQ213752.1 | - | 5 - 6 | 517 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |