Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009656.1 | NP_001009656.1 | 1 - 2 | 703 | 86 | |
XM_006235749.1 | XP_006235811.1 | 2 - 3 | 699 | 86 | ||
XM_006235750.1 | XP_006235812.1 | 2 - 3 | 664 | 86 | ||
XM_006235751.2 | XP_006235813.1 | 2 - 3 | 685 | 86 | ||
XM_006235752.2 | XP_006235814.1 | 1 - 2 | 599 | 86 | ||
GenBank mRNA | BC087720.1 | - | 1 - 2 | 703 | 7318724 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |