Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214459.2 | NP_999624.1 | 6 - 7 | 1059 | 73 | |
GenBank mRNA | AY500157.1 | - | 6 - 7 | 1059 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |