Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031803.1 | NP_113991.1 | 7 - 8 | 803 | 117 | |
GenBank mRNA | AF059273.1 | - | 7 - 8 | 803 | 117 | |
AF205778.1 | - | 697 | 117 | |||
AF205779.1 | - | 697 | 117 | |||
AF205780.1 | - | 697 | 117 | |||
FQ211504.1 | - | 847 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |