Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2w1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248980.2 | XP_006249042.1 | 1 - 2 | 182 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |