Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248842.2 | XP_006248904.1 | 15 - 16 | 2544 | 74 | |
XM_006248844.2 | XP_006248906.1 | 15 - 16 | 2544 | 74 | ||
XM_346914.7 | XP_346915.4 | 14 - 15 | 2381 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |