Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006248842.2 | XP_006248904.1 | 19 - 20 | 3018 | 86 | |
XM_346914.7 | XP_346915.4 | 18 - 19 | 2855 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |