Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108860.1 | NP_001102330.1 | 9 - 10 | 1021 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |