Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271181.1 | NP_001258110.1 | 3 - 4 | 265 | 71 | |
GenBank mRNA | FM129783.1 | - | 4 - 5 | 280 | 71 | |
FM139638.1 | - | 3 - 4 | 265 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |