Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271181.1 | NP_001258110.1 | 5 - 6 | 364 | 85 | |
GenBank mRNA | FM129783.1 | - | 6 - 7 | 379 | 85 | |
FM139638.1 | - | 5 - 6 | 84 | |||
FQ214213.1 | - | 5 - 6 | 353 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |