Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_573585.6 | XP_573585.3 | 2 - 3 | 467 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |