Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC058444.1 | - | 3 - 4 | 382 | 6918440 | 86 |
FQ225009.1 | - | 3 - 4 | 381 | 86 | ||
FQ225217.1 | - | 3 - 4 | 381 | 86 | ||
FQ225221.1 | - | 3 - 4 | 381 | 86 | ||
FQ225582.1 | - | 3 - 4 | 378 | 86 | ||
FQ225670.1 | - | 3 - 4 | 381 | 86 | ||
FQ226479.1 | - | 3 - 4 | 378 | 86 | ||
FQ231406.1 | - | 3 - 4 | 381 | 86 | ||
FQ232883.1 | - | 3 - 4 | 381 | 86 | ||
FQ233623.1 | - | 3 - 4 | 381 | 86 | ||
FQ233884.1 | - | 3 - 4 | 381 | 86 | ||
U22520.1 | - | 3 - 4 | 366 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |