Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006958.1 | NP_001006959.1 | 14 - 15 | 1247 | 91 | |
XM_006250722.2 | XP_006250784.1 | 14 - 15 | 1317 | 91 | ||
GenBank mRNA | BC083620.1 | - | 14 - 15 | 1247 | 7133993 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |