Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100890.1 | NP_001094360.1 | 6 - 7 | 1144 | 71 | |
XM_006250775.2 | XP_006250837.1 | 6 - 7 | 1031 | 71 | ||
GenBank mRNA | BC103654.1 | - | 1 - 2 | 115 | 71 | |
CK601425.1 | - | 4 - 5 | 560 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |