Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rchy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007618.1 | NP_001007619.1 | 3 - 4 | 714 | 137 | |
GenBank mRNA | BC083739.1 | - | 3 - 4 | 714 | 7191744 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |