Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013988.1 | NP_001014010.1 | 1 - 2 | 284 | 162 | |
GenBank mRNA | BC089867.1 | - | 1 - 2 | 284 | 7321620 | 162 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |