Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100889.1 | NP_001094359.1 | 4 - 5 | 578 | 68 | |
XM_006250723.2 | XP_006250785.1 | 4 - 5 | 1589 | 68 | ||
XM_006250725.2 | XP_006250787.1 | 2 - 3 | 3362 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |