Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100889.1 | NP_001094359.1 | 8 - 9 | 5169 | 119 | |
XM_006250723.2 | XP_006250785.1 | 8 - 9 | 6183 | 119 | ||
XM_006250725.2 | XP_006250787.1 | 6 - 7 | 7956 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |