Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172320.1 | NP_758823.1 | 11 - 12 | 1424 | 111 | |
XM_008770001.1 | XP_008768223.1 | 11 - 12 | 1472 | 111 | ||
GenBank mRNA | BC091391.1 | - | 8 - 9 | 989 | 111 | |
FQ213773.1 | - | 7 - 8 | 860 | 111 | ||
FQ219466.1 | - | 8 - 9 | 958 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |