Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105998.1 | NP_001099468.1 | 5 - 6 | 798 | 149 | |
XM_006250715.2 | XP_006250777.1 | 4 - 5 | 898 | 149 | ||
GenBank mRNA | FQ223715.1 | - | 4 - 5 | 891 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Probe spans exons |