Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024158.1 | NP_077072.1 | 4 - 5 | 574 | 99 | |
GenBank mRNA | FQ212366.1 | - | 879 | 99 | ||
L33894.1 | - | 4 - 5 | 574 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |