Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Areg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017123.1 | NP_058819.1 | 2 - 3 | 459 | 119 | |
GenBank mRNA | X55183.1 | - | 2 - 3 | 459 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |