Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam47e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770095.1 | XP_008768317.1 | 5 - 6 | 960 | 84 | |
XM_008770096.1 | XP_008768318.1 | 4 - 5 | 867 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |