Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100510.1 | NP_001093980.1 | 5 - 6 | 649 | 63 | |
GenBank mRNA | BC158592.1 | - | 5 - 6 | 649 | 8362956 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |