Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008770650.1 | XP_008768872.1 | 3 - 4 | 755 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |