Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025141.1 | NP_001020312.1 | 5 - 6 | 1047 | 129 | |
XM_006251901.2 | XP_006251963.1 | 6 - 7 | 1457 | 129 | ||
XM_006251903.2 | XP_006251965.1 | 6 - 7 | 1753 | 129 | ||
XM_006251904.1 | XP_006251966.1 | 4 - 5 | 934 | 129 | ||
XM_008770563.1 | XP_008768785.1 | 6 - 7 | 1908 | 129 | ||
XM_008770564.1 | XP_008768786.1 | 7 - 8 | 1805 | 129 | ||
XM_008770565.1 | XP_008768787.1 | 7 - 8 | 1678 | 129 | ||
XM_008770566.1 | XP_008768788.1 | 6 - 7 | 1739 | 129 | ||
XM_008770567.1 | XP_008768789.1 | 5 - 6 | 1560 | 129 | ||
XM_008770568.1 | XP_008768790.1 | 6 - 7 | 2316 | 129 | ||
GenBank mRNA | BC091423.1 | - | 5 - 6 | 1047 | 7365771 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |