Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251872.2 | XP_006251934.1 | 1 | 300 | 76 | |
XM_008770517.1 | XP_008768739.1 | 2 | 145 | 76 | ||
XM_008770518.1 | XP_008768740.1 | 2 | 119 | 76 |
Gene Symbol: LOC103694852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773788.1 | XP_008772010.1 | 1 | 300 | 76 | |
XM_008773789.1 | XP_008772011.1 | 2 | 148 | 76 | ||
XM_008773790.1 | XP_008772012.1 | 2 | 122 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |