Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109082.1 | NP_001102552.1 | 4 - 5 | 632 | 79 | |
XM_006250463.2 | XP_006250525.1 | 4 - 5 | 647 | 79 | ||
XM_006250465.2 | XP_006250527.1 | 4 - 5 | 647 | 79 | ||
XM_008769868.1 | XP_008768090.1 | 4 - 5 | 647 | 79 | ||
XM_008769869.1 | XP_008768091.1 | 4 - 5 | 970 | 79 | ||
XM_008769870.1 | XP_008768092.1 | 4 - 5 | 647 | 79 | ||
XM_008769871.1 | XP_008768093.1 | 4 - 5 | 647 | 79 | ||
XM_008769872.1 | XP_008768094.1 | 4 - 5 | 647 | 79 | ||
GenBank mRNA | AY724511.1 | - | 4 - 5 | 592 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |