Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adck3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013185.1 | NP_001013203.1 | 4 - 5 | 694 | 65 | |
XM_006250387.1 | XP_006250449.1 | 4 - 5 | 777 | 65 | ||
GenBank mRNA | BC091388.1 | - | 4 - 5 | 694 | 7321446 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |