Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769812.1 | XP_008768034.1 | 9 - 10 | 1892 | 119 | |
GenBank mRNA | AB038495.1 | - | 5 - 6 | 1749 | 119 |
Gene Symbol: LOC100909664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751234.3 | XP_003751282.2 | 9 - 10 | 2787 | 119 | |
XM_006249680.2 | XP_006249742.1 | 8 - 9 | 2974 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |