Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012724.4 | NP_036856.1 | 3 - 4 | 638 | 116 | |
XM_008769780.1 | XP_008768002.1 | 2 - 3 | 1343 | 116 | ||
XM_008769781.1 | XP_008768003.1 | 2 - 3 | 1343 | 116 | ||
GenBank mRNA | J03606.1 | - | 3 - 4 | 638 | 116 | |
M17153.1 | - | 3 - 4 | 642 | 116 | ||
M21622.1 | - | 3 - 4 | 701 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons |