Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106521.2 | NP_001099991.1 | 1 - 2 | 107 | 133 | |
GenBank mRNA | BC167042.1 | - | 1 - 2 | 107 | 7444530 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Probe spans exons |