Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108967.1 | NP_001102437.1 | 2 - 3 | 1397 | 99 | |
XM_006235788.1 | XP_006235850.1 | 2 - 3 | 1397 | 99 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Rattus norvegicus Probe spans exons |