Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108254.1 | NP_001101724.1 | 6 - 7 | 849 | 148 | |
XM_008773171.1 | XP_008771393.1 | 6 - 7 | 889 | 148 | ||
XM_008773172.1 | XP_008771394.1 | 6 - 7 | 889 | 148 | ||
GenBank mRNA | BC081944.1 | - | 6 - 7 | 818 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |