Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773127.1 | XP_008771349.1 | 1 - 2 | 579 | 108 | |
GenBank mRNA | CB586797.1 | - | 1 - 2 | 299 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel Rattus norvegicus Probe spans exons |