Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022213.1 | NP_071549.1 | 6 - 7 | 765 | 83 | |
XM_006238714.2 | XP_006238776.1 | 7 - 8 | 786 | 83 | ||
XM_008763979.1 | XP_008762201.1 | 7 - 8 | 988 | 83 | ||
GenBank mRNA | D64047.1 | - | 6 - 7 | 765 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |