Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004236.1 | NP_001004236.1 | 7 - 8 | 813 | 104 | |
XM_006238639.1 | XP_006238701.1 | 9 - 10 | 1086 | 104 | ||
GenBank mRNA | BC078938.1 | - | 7 - 8 | 813 | 7108356 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |