Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133396.2 | NP_596887.1 | 5 - 6 | 889 | 115 | |
XM_006238628.2 | XP_006238690.1 | 5 - 6 | 1120 | 115 | ||
GenBank mRNA | AB049402.1 | - | 5 - 6 | 890 | 115 | |
BC081737.1 | - | 6 - 7 | 1140 | 115 | ||
BC128727.1 | - | 5 - 6 | 895 | 7111312 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Rattus norvegicus Probe spans exons |