Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038992.1 | NP_001034081.1 | 1 - 2 | 99 | 84 | |
GenBank mRNA | BC099226.1 | - | 1 - 2 | 99 | 7388643 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |