Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106675.1 | NP_001100145.1 | 6 - 7 | 586 | 98 | |
XM_006238507.1 | XP_006238569.1 | 4 - 5 | 517 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |