Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106680.1 | NP_001100150.1 | 7 - 8 | 1043 | 153 | |
XM_006238651.1 | XP_006238713.1 | 7 - 8 | 1038 | 153 | ||
XM_006238652.2 | XP_006238714.1 | 6 - 7 | 1368 | 153 | ||
XM_006238653.2 | XP_006238715.1 | 6 - 7 | 1290 | 153 | ||
XM_006238654.2 | XP_006238716.1 | 6 - 7 | 1391 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |