Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134626.1 | NP_001128098.1 | 1 - 2 | 222 | 53 | |
XM_006238710.2 | XP_006238772.1 | 1 - 2 | 421 | 53 | ||
XM_008764053.1 | XP_008762275.1 | 1 - 2 | 421 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |