Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013118.1 | NP_037250.1 | 2 - 3 | 291 | 76 | |
GenBank mRNA | M93005.1 | - | 2 - 3 | 291 | 76 | |
M95493.1 | - | 2 - 3 | 291 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |