Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047093.1 | NP_001040558.1 | 7 - 8 | 1694 | 72 | |
XM_006250959.2 | XP_006251021.1 | 7 - 8 | 1350 | 72 | ||
XM_008770157.1 | XP_008768379.1 | 7 - 8 | 1172 | 72 | ||
GenBank mRNA | AY641809.1 | - | 7 - 8 | 1694 | 72 | |
BC099156.1 | - | 1 - 2 | 105 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |