Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191068.1 | NP_001177997.1 | 4 - 5 | 646 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |