Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108137.2 | NP_001101607.1 | 6 - 7 | 937 | 67 | |
XM_006242933.2 | XP_006242995.1 | 6 - 7 | 1382 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |