Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108762.1 | NP_001102232.1 | 5 - 6 | 344 | 141 | |
XM_006243133.2 | XP_006243195.1 | 6 - 7 | 470 | 141 | ||
GenBank mRNA | FQ213793.1 | - | 5 - 6 | 610 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |