Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012540.2 | NP_036672.2 | 3 - 4 | 1037 | 95 | |
XM_006243150.2 | XP_006243212.1 | 3 - 4 | 1034 | 95 | ||
GenBank mRNA | X00469.1 | - | 3 - 4 | 1037 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |