Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stoml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243184.2 | XP_006243246.1 | 3 - 4 | 525 | 72 | |
XM_236297.8 | XP_236297.3 | 4 - 5 | 675 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |